The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006657.1 | NP_001006658.1 | 18 - 19 | 2070 | 159 |
NM_020779.3 | NP_065830.2 | 17 - 18 | 2037 | 159 | |
XM_011533007.2 | XP_011531309.1 | 7 - 8 | 762 | 159 | |
GenBank mRNA | AB037757.2 | - | 18 - 19 | 2078 | 159 |
AK091205.1 | - | 11 - 12 | 1405 | 159 | |
AK122917.1 | - | 17 - 18 | 1962 | 159 | |
AK315094.1 | - | 17 - 18 | 1962 | 159 | |
AL832903.1 | - | 6 - 7 | 567 | 159 | |
BC036659.2 | - | 17 - 18 | 2036 | 159 |