The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144885.1 | NP_001138357.1 | 2 - 3 | 213 | 109 |
XM_011530958.1 | XP_011529260.1 | 2 - 3 | 204 | 109 | |
GenBank mRNA | N27803.1 | - | 2 - 3 | 191 | 109 |