Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.9: 111686175 - 111795949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_109816.1 3 - 4 319 129
XM_006716974.2 XP_006717037.1 3 - 4 303 129
XM_006716975.2 XP_006717038.1 3 - 4 303 129
XM_011518309.1 XP_011516611.1 3 - 4 303 129
XM_017014340.1 XP_016869829.1 3 - 4 1260 129
XM_017014341.1 XP_016869830.1 3 - 4 303 129
GenBank mRNA AK057341.1 - 3 - 4 254 129
AL704322.1 - 3 - 4 320 129
BC112357.1 - 3 - 4 247 129
BC144680.1 - 3 - 4 243 129

View Full Product Details