Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.9: 111686175 - 111795949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080551.2 NP_001074020.2 2 - 3 538 152
NM_173521.4 NP_775792.4 4 - 5 545 152
NR_109816.1 6 - 7 747 152
XM_006716974.2 XP_006717037.1 6 - 7 731 152
XM_006716975.2 XP_006717038.1 6 - 7 731 152
XM_011518302.2 XP_011516604.1 4 - 5 2540 152
XM_011518303.1 XP_011516605.1 4 - 5 512 152
XM_011518309.1 XP_011516611.1 6 - 7 731 152
XM_017014341.1 XP_016869830.1 6 - 7 731 152
GenBank mRNA AK057341.1 - 6 - 7 682 152
AK126055.1 - 2 - 3 538 152
AL833535.1 - 4 - 5 539 152
BC112357.1 - 6 - 7 675 152
BC144680.1 - 6 - 7 671 152

View Full Product Details