Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LETM2
Entrez Gene ID
137994
Gene Name
leucine zipper and EF-hand containing transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.8: 38382505 - 38409527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286787.1 NP_001273716.1 2 - 3 210 121
NM_001286819.1 NP_001273748.1 2 - 3 210 121
NM_001286821.1 NP_001273750.1 2 - 3 210 121
XM_011544408.2 XP_011542710.1 2 - 3 2706 121
XM_011544409.2 XP_011542711.1 2 - 3 256 121
XM_017013057.1 XP_016868546.1 2 - 3 2704 121
GenBank mRNA AK289944.1 - 2 - 3 170 121
AL832241.1 - 2 - 3 141 121
BC069830.1 - 2 - 3 144 121

View Full Product Details