The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053004.2 | NP_443730.1 | 2 - 3 | 218 | 102 |
GenBank mRNA | AB051432.1 | - | 2 - 3 | 159 | 102 |
AF238328.1 | - | 2 - 3 | 218 | 102 | |
AF301895.1 | - | 2 - 3 | 183 | 102 | |
AY007378.1 | - | 2 - 3 | 218 | 102 | |
BC012060.1 | - | 2 - 3 | 119 | 102 |