Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TCF12
- Entrez Gene ID
-
6938
- Gene Name
- transcription factor 12
- Gene Aliases
-
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
- Chromosome Location
-
Chr.15: 56918090 - 57291261 on Build GRCh38
View Full Product Details