The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025238.3 | NP_079514.1 | 6 - 7 | 1341 | 80 |
GenBank mRNA | AF257241.1 | - | 6 - 7 | 1220 | 80 |
AF355402.1 | - | 6 - 7 | 1221 | 80 | |
AF529369.1 | - | 6 - 7 | 1138 | 80 | |
AK000731.1 | - | 6 - 7 | 1205 | 80 | |
AK225355.1 | - | 6 - 7 | 1205 | 80 | |
AK225362.1 | - | 6 - 7 | 1205 | 80 | |
AK298534.1 | - | 3 - 4 | 488 | 80 | |
AL136853.1 | - | 6 - 7 | 1221 | 80 | |
AY364253.1 | - | 6 - 7 | 1138 | 80 | |
BC017464.1 | - | 6 - 7 | 1168 | 80 | |
BC028097.1 | - | 6 - 7 | 1241 | 80 |