The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018590.4 | NP_061060.3 | 5 - 6 | 1532 | 122 |
NR_135077.1 | 6 - 7 | 1693 | 122 | ||
XM_011539938.1 | XP_011538240.1 | 5 - 6 | 1532 | 122 | |
GenBank mRNA | AB079252.1 | - | 5 - 6 | 1463 | 122 |
AB090811.1 | - | 5 - 6 | 1491 | 122 | |
AK074474.1 | - | 1 - 2 | 184 | 122 | |
AK125300.1 | - | 5 - 6 | 1444 | 122 | |
BC030268.1 | - | 5 - 6 | 1491 | 122 | |
BX640967.1 | - | 6 - 7 | 1589 | 122 | |
BX641073.1 | - | 1 - 2 | 1589 | 122 | |
KU178704.1 | - | 4 - 5 | 1156 | 122 |