Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 23 - 24 2923 74
NM_001122634.3 NP_001116106.1 12 - 13 1979 74
NM_001875.4 NP_001866.2 22 - 23 2958 74
XM_011510640.1 XP_011508942.1 24 - 25 3157 74
XM_011510641.2 XP_011508943.1 24 - 25 3301 74
XM_011510642.2 XP_011508944.1 23 - 24 3173 74
XM_011510643.2 XP_011508945.1 24 - 25 3038 74
XM_011510644.2 XP_011508946.1 23 - 24 3079 74
GenBank mRNA AB180934.1 - 23 - 24 2823 74
AB180935.1 - 23 - 24 2823 74
AB180936.1 - 23 - 24 2823 74
AB180938.1 - 23 - 24 2823 74
AB208800.1 - 24 - 25 2899 74
AF154830.1 - 23 - 24 2946 74
AK289749.1 - 23 - 24 2958 74
AK302778.1 - 13 - 14 1968 74
AY317138.1 - 12 - 13 1965 74
BC140943.1 - 22 - 23 2925 74
BX640601.1 - 4 - 5 443 74
D90282.1 - 22 - 23 2941 74
Y15793.1 - 22 - 23 2823 74

View Full Product Details