Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 28 - 29 3501 106
NM_001122634.3 NP_001116106.1 17 - 18 2557 106
NM_001875.4 NP_001866.2 27 - 28 3536 106
XM_011510640.1 XP_011508942.1 29 - 30 3735 106
XM_011510641.2 XP_011508943.1 29 - 30 3879 106
XM_011510642.2 XP_011508944.1 28 - 29 3751 106
XM_011510643.2 XP_011508945.1 29 - 30 3616 106
XM_011510644.2 XP_011508946.1 28 - 29 3657 106
GenBank mRNA AB180934.1 - 28 - 29 3401 106
AB180935.1 - 28 - 29 3401 106
AB180936.1 - 28 - 29 3401 106
AB180938.1 - 28 - 29 3401 106
AB208800.1 - 29 - 30 3477 106
AF154830.1 - 28 - 29 3524 106
AK289749.1 - 28 - 29 3536 106
AK302778.1 - 18 - 19 2546 106
AY317138.1 - 17 - 18 2543 106
BC140943.1 - 27 - 28 3503 106
BX640601.1 - 9 - 10 1021 106
D90282.1 - 27 - 28 3519 106
Y15793.1 - 27 - 28 3401 106

View Full Product Details