Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 35 - 36 4204 65
NM_001122634.3 NP_001116106.1 24 - 25 3260 65
NM_001875.4 NP_001866.2 34 - 35 4239 65
XM_011510640.1 XP_011508942.1 36 - 37 4438 65
XM_011510641.2 XP_011508943.1 36 - 37 4582 65
XM_011510642.2 XP_011508944.1 35 - 36 4454 65
XM_011510643.2 XP_011508945.1 36 - 37 4319 65
XM_011510644.2 XP_011508946.1 35 - 36 4360 65
GenBank mRNA AB180934.1 - 35 - 36 4104 65
AB180936.1 - 35 - 36 4104 65
AB180938.1 - 35 - 36 4104 65
AB208800.1 - 36 - 37 4180 65
AF154830.1 - 35 - 36 4227 65
AK289749.1 - 35 - 36 4239 65
AK302778.1 - 25 - 26 3249 65
AY317138.1 - 24 - 25 3246 65
BC140943.1 - 34 - 35 4206 65
BX640601.1 - 16 - 17 1724 65
D90282.1 - 34 - 35 4222 65
Y15793.1 - 34 - 35 4104 65

View Full Product Details