Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 36 - 37 4256 77
NM_001122634.3 NP_001116106.1 25 - 26 3312 77
NM_001875.4 NP_001866.2 35 - 36 4291 77
XM_011510640.1 XP_011508942.1 37 - 38 4490 77
XM_011510641.2 XP_011508943.1 37 - 38 4634 77
XM_011510642.2 XP_011508944.1 36 - 37 4506 77
XM_011510643.2 XP_011508945.1 37 - 38 4371 77
XM_011510644.2 XP_011508946.1 36 - 37 4412 77
GenBank mRNA AB180934.1 - 36 - 37 4156 77
AB180936.1 - 36 - 37 4156 77
AB180938.1 - 36 - 37 4156 77
AB208800.1 - 37 - 38 4232 77
AF154830.1 - 36 - 37 4279 77
AK289749.1 - 36 - 37 4291 77
AK302778.1 - 26 - 27 3301 77
AY317138.1 - 25 - 26 3298 77
BC140943.1 - 35 - 36 4258 77
BX537776.1 - 1 - 2 3344 77
BX640601.1 - 17 - 18 1776 77
D90282.1 - 35 - 36 4274 77
Y15793.1 - 35 - 36 4156 77

View Full Product Details