Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
Chromosome Location
Chr.2: 210477682 - 210679107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122633.2 NP_001116105.1 37 - 38 4376 123
NM_001122634.3 NP_001116106.1 26 - 27 3432 123
NM_001875.4 NP_001866.2 36 - 37 4411 123
XM_011510640.1 XP_011508942.1 38 - 39 4610 123
XM_011510641.2 XP_011508943.1 38 - 39 4754 123
XM_011510642.2 XP_011508944.1 37 - 38 4626 123
XM_011510643.2 XP_011508945.1 38 - 39 4491 123
XM_011510644.2 XP_011508946.1 37 - 38 4532 123
GenBank mRNA AB180934.1 - 37 - 38 4276 123
AB180936.1 - 37 - 38 4276 123
AB180938.1 - 37 - 38 4276 123
AB208800.1 - 38 - 39 4352 123
AF154830.1 - 37 - 38 4399 123
AK289749.1 - 37 - 38 4411 123
AK302778.1 - 27 - 28 3421 123
AY317138.1 - 26 - 27 3418 123
BC140943.1 - 36 - 37 4378 123
BX537776.1 - 2 - 3 3464 123
BX640601.1 - 18 - 19 1896 123
D90282.1 - 36 - 37 4394 123
Y15793.1 - 36 - 37 4276 123

View Full Product Details