The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271466.2 | NP_001258395.1 | 22 - 23 | 2299 | 57 |
XM_011511905.1 | XP_011510207.1 | 22 - 23 | 2256 | 57 | |
XM_011511906.1 | XP_011510208.1 | 22 - 23 | 2253 | 57 | |
XM_011511907.1 | XP_011510209.1 | 21 - 22 | 2157 | 57 | |
XM_011511910.1 | XP_011510212.1 | 23 - 24 | 2372 | 57 | |
XM_011511911.1 | XP_011510213.1 | 23 - 24 | 2372 | 57 | |
XM_011511912.1 | XP_011510214.1 | 22 - 23 | 2256 | 57 | |
XM_011511919.2 | XP_011510221.1 | 12 - 13 | 1644 | 57 | |
XM_017005018.1 | XP_016860507.1 | 22 - 23 | 2514 | 57 | |
XM_017005019.1 | XP_016860508.1 | 22 - 23 | 2432 | 57 | |
XM_017005021.1 | XP_016860510.1 | 23 - 24 | 2372 | 57 | |
XM_017005023.1 | XP_016860512.1 | 23 - 24 | 2630 | 57 | |
XM_017005025.1 | XP_016860514.1 | 22 - 23 | 2256 | 57 | |
GenBank mRNA | AB058771.1 | - | 12 - 13 | 1195 | 57 |