Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKAG3
Entrez Gene ID
53632
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 3
Gene Aliases
AMPKG3
Chromosome Location
Chr.2: 218821853 - 218831803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017431.2 NP_059127.2 13 - 14 94
GenBank mRNA AJ249977.1 - 13 - 14 1608 95
AK300078.1 - 14 - 15 94

View Full Product Details