The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128911.1 | NP_001122383.1 | 12 - 13 | 1189 | 82 |
NM_001128912.1 | NP_001122384.1 | 12 - 13 | 1177 | 82 | |
GenBank mRNA | AB208825.1 | - | 11 - 12 | 911 | 82 |
AK299210.1 | - | 8 - 9 | 648 | 82 | |
X78136.1 | - | 11 - 12 | 861 | 82 |