The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138290.2 | NP_612147.1 | 2 - 3 | 642 | 126 |
XM_011515826.1 | XP_011514128.1 | 2 - 3 | 642 | 126 | |
XM_011515827.1 | XP_011514129.1 | 2 - 3 | 642 | 126 | |
XM_017011761.1 | XP_016867250.1 | 2 - 3 | 251 | 126 | |
XM_017011762.1 | XP_016867251.1 | 3 - 4 | 176 | 126 | |
XM_017011766.1 | XP_016867255.1 | 2 - 3 | 442 | 126 | |
GenBank mRNA | AK055233.1 | - | 2 - 3 | 640 | 126 |