The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_134959.1 | 8 - 9 | 1141 | 74 | |
GenBank mRNA | AJ606321.1 | - | 8 - 9 | 1141 | 74 |
AY787459.1 | - | 8 - 9 | 942 | 74 | |
AY787460.1 | - | 8 - 9 | 942 | 74 |