The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007098.2 | NP_001007099.1 | 4 - 5 | 428 | 84 |
NM_001193599.1 | NP_001180528.1 | 4 - 5 | 488 | 84 | |
NM_001193600.1 | NP_001180529.1 | 4 - 5 | 428 | 84 | |
NM_001193617.1 | NP_001180546.1 | 4 - 5 | 502 | 84 | |
NM_002979.4 | NP_002970.2 | 5 - 6 | 560 | 84 | |
XM_005271103.4 | XP_005271160.1 | 5 - 6 | 503 | 84 | |
XM_011541935.2 | XP_011540237.1 | 5 - 6 | 503 | 84 | |
XM_017002045.1 | XP_016857534.1 | 5 - 6 | 503 | 84 | |
GenBank mRNA | AB208789.1 | - | 5 - 6 | 512 | 84 |
AK294631.1 | - | 4 - 5 | 431 | 84 | |
AK295214.1 | - | 4 - 5 | 445 | 84 | |
AK312856.1 | - | 5 - 6 | 511 | 84 | |
BC067108.1 | - | 4 - 5 | 339 | 84 | |
M75883.1 | - | 5 - 6 | 413 | 84 |