The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160.2 | NP_001151.1 | 20 - 21 | 3370 | 105 |
NM_013229.2 | NP_037361.1 | 21 - 22 | 3499 | 105 | |
NM_181861.1 | NP_863651.1 | 21 - 22 | 3532 | 105 | |
NM_181868.1 | NP_863658.1 | 20 - 21 | 3403 | 105 | |
GenBank mRNA | AB007873.2 | - | 20 - 21 | 2922 | 105 |
AF013263.1 | - | 20 - 21 | 3370 | 105 | |
AF134397.1 | - | 20 - 21 | 2922 | 105 | |
AF149794.1 | - | 20 - 21 | 2955 | 105 | |
AJ243003.1 | - | 19 - 20 | 2826 | 105 | |
AJ243004.1 | - | 19 - 20 | 2826 | 105 | |
AJ243005.1 | - | 19 - 20 | 2826 | 105 | |
AJ243006.1 | - | 19 - 20 | 2826 | 105 | |
AJ243007.1 | - | 19 - 20 | 2826 | 105 | |
AJ243008.1 | - | 19 - 20 | 2826 | 105 | |
AJ243009.1 | - | 20 - 21 | 2955 | 105 | |
AJ243010.1 | - | 19 - 20 | 2826 | 105 | |
AJ243011.1 | - | 19 - 20 | 2826 | 105 | |
AJ243048.1 | - | 20 - 21 | 2955 | 105 | |
AJ243107.1 | - | 19 - 20 | 2850 | 105 | |
BC136531.1 | - | 21 - 22 | 3103 | 105 | |
BC136532.1 | - | 21 - 22 | 3103 | 105 | |
BC150255.1 | - | 20 - 21 | 2951 | 105 | |
EF560718.1 | - | 19 - 20 | 2831 | 105 |