Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBPL3
Entrez Gene ID
26031
Gene Name
oxysterol binding protein like 3
Gene Aliases
ORP-3, ORP3, OSBP3
Chromosome Location
Chr.7: 24796537 - 24980218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015550.3 NP_056365.1 14 - 15 1972 146
NM_145320.2 NP_663160.1 12 - 13 1552 146
NM_145321.2 NP_663161.1 12 - 13 1537 146
NM_145322.2 NP_663162.1 11 - 12 1444 146
NR_104111.1 13 - 14 1645 146
NR_104112.1 12 - 13 1552 146
XM_005249698.3 XP_005249755.1 14 - 15 1692 146
XM_006715681.3 XP_006715744.1 13 - 14 1866 146
XM_006715682.3 XP_006715745.1 13 - 14 1870 146
XM_006715683.3 XP_006715746.1 12 - 13 1777 146
XM_006715684.3 XP_006715747.1 13 - 14 1629 146
XM_011515258.2 XP_011513560.1 14 - 15 1769 146
XM_011515259.2 XP_011513561.1 15 - 16 2266 146
XM_017011948.1 XP_016867437.1 16 - 17 2129 146
XM_017011949.1 XP_016867438.1 14 - 15 2158 146
GenBank mRNA AB014604.2 - 14 - 15 1909 146
AF392444.1 - 14 - 15 1881 146
AF491781.1 - 13 - 14 1599 146
AF491782.1 - 12 - 13 1506 146
AF491783.1 - 12 - 13 1491 146
AF491784.1 - 11 - 12 1398 146
AF491785.1 - 13 - 14 1599 146
AF491786.1 - 12 - 13 1506 146
AF515639.1 - 6 - 7 651 146
AF515640.1 - 5 - 6 558 146
AL832293.1 - 11 - 12 1208 146
AY008372.1 - 14 - 15 1802 146
BC017731.1 - 14 - 15 1901 146

View Full Product Details