The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015550.3 | NP_056365.1 | 19 - 20 | 2653 | 83 |
NM_145320.2 | NP_663160.1 | 17 - 18 | 2233 | 83 | |
NM_145321.2 | NP_663161.1 | 17 - 18 | 2218 | 83 | |
NM_145322.2 | NP_663162.1 | 16 - 17 | 2125 | 83 | |
NR_104111.1 | 18 - 19 | 2725 | 83 | ||
NR_104112.1 | 17 - 18 | 2632 | 83 | ||
XM_005249698.3 | XP_005249755.1 | 19 - 20 | 2373 | 83 | |
XM_006715681.3 | XP_006715744.1 | 18 - 19 | 2547 | 83 | |
XM_006715682.3 | XP_006715745.1 | 18 - 19 | 2551 | 83 | |
XM_006715683.3 | XP_006715746.1 | 17 - 18 | 2458 | 83 | |
XM_006715684.3 | XP_006715747.1 | 18 - 19 | 2310 | 83 | |
XM_011515258.2 | XP_011513560.1 | 19 - 20 | 2450 | 83 | |
XM_011515259.2 | XP_011513561.1 | 20 - 21 | 2947 | 83 | |
XM_017011948.1 | XP_016867437.1 | 21 - 22 | 2810 | 83 | |
XM_017011949.1 | XP_016867438.1 | 19 - 20 | 2839 | 83 | |
GenBank mRNA | AB014604.2 | - | 19 - 20 | 2590 | 83 |
AF392444.1 | - | 19 - 20 | 2562 | 83 | |
AF491781.1 | - | 18 - 19 | 2280 | 83 | |
AF491782.1 | - | 17 - 18 | 2187 | 83 | |
AF491783.1 | - | 17 - 18 | 2172 | 83 | |
AF491784.1 | - | 16 - 17 | 2079 | 83 | |
AF491785.1 | - | 18 - 19 | 2679 | 83 | |
AF491786.1 | - | 17 - 18 | 2586 | 83 | |
AL832293.1 | - | 16 - 17 | 1889 | 83 | |
AY008372.1 | - | 19 - 20 | 2483 | 83 | |
BC017731.1 | - | 19 - 20 | 2582 | 83 |