Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBTPS1
Entrez Gene ID
8720
Gene Name
membrane bound transcription factor peptidase, site 1
Gene Aliases
PCSK8, S1P, SKI-1
Chromosome Location
Chr.16: 84053761 - 84116943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003791.3 NP_003782.1 13 - 14 2313 72
XM_017023816.1 XP_016879305.1 12 - 13 2062 72
XM_017023817.1 XP_016879306.1 14 - 15 2316 72
GenBank mRNA AK091212.1 - 2 - 3 1024 72
AK291773.1 - 13 - 14 2196 72
AK299067.1 - 12 - 13 1538 72
BC114555.1 - 13 - 14 2199 72
BC114961.1 - 13 - 14 2199 72
D42053.1 - 13 - 14 2276 72

View Full Product Details