The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006767.3 | NP_006758.2 | 13 - 14 | 1551 | 116 |
GenBank mRNA | AK296347.1 | - | 12 - 13 | 1432 | 116 |
AK310747.1 | - | 11 - 12 | 1154 | 116 | |
AK313502.1 | - | 8 - 9 | 953 | 116 | |
BC026214.1 | - | 13 - 14 | 1453 | 116 | |
CT841521.1 | - | 13 - 14 | 1467 | 116 | |
D38496.1 | - | 13 - 14 | 1446 | 116 |