The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006767.3 | NP_006758.2 | 1 - 2 | 303 | 61 |
GenBank mRNA | AK296347.1 | - | 1 - 2 | 241 | 61 |
BC026214.1 | - | 1 - 2 | 61 | ||
CT841521.1 | - | 1 - 2 | 219 | 61 | |
D38496.1 | - | 1 - 2 | 60 | ||
DA194822.1 | - | 1 - 2 | 303 | 61 |