Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A11
Entrez Gene ID
23657
Gene Name
solute carrier family 7 member 11
Gene Aliases
CCBR1, xCT
Chromosome Location
Chr.4: 138164094 - 138242418 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014331.3 NP_055146.1 10 - 11 1549 72
XM_011531802.2 XP_011530104.1 10 - 11 1618 72
GenBank mRNA AB026891.1 - 10 - 11 1504 72
AB040875.1 - 10 - 11 1404 72
AF200708.1 - 10 - 11 1514 72
AF252872.1 - 10 - 11 1500 72
AJ277882.1 - 10 - 11 1269 72
AK290359.1 - 10 - 11 1549 72
AK314855.1 - 10 - 11 1443 72
BC012087.1 - 10 - 11 1429 72

View Full Product Details