The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014331.3 | NP_055146.1 | 1 - 2 | 555 | 142 |
XM_011531802.2 | XP_011530104.1 | 1 - 2 | 624 | 142 | |
GenBank mRNA | AB026891.1 | - | 1 - 2 | 510 | 142 |
AB040875.1 | - | 1 - 2 | 410 | 142 | |
AF200708.1 | - | 1 - 2 | 520 | 142 | |
AJ277882.1 | - | 1 - 2 | 275 | 142 | |
AK290359.1 | - | 1 - 2 | 555 | 142 | |
AK314855.1 | - | 1 - 2 | 449 | 142 | |
BC012087.1 | - | 1 - 2 | 435 | 142 |