The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001814.5 | NP_001805.3 | 6 - 7 | 1000 | 83 |
GenBank mRNA | AF234263.1 | - | 6 - 7 | 911 | 83 |
AF234264.1 | - | 6 - 7 | 911 | 83 | |
AF254757.1 | - | 6 - 7 | 911 | 83 | |
AF525033.1 | - | 6 - 7 | 911 | 83 | |
AK223038.1 | - | 6 - 7 | 949 | 83 | |
AK292117.1 | - | 6 - 7 | 918 | 83 | |
AK296190.1 | - | 7 - 8 | 898 | 83 | |
X87212.1 | - | 6 - 7 | 918 | 83 |