The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003952.2 | NP_003943.2 | 3 - 4 | 323 | 124 |
XM_005274164.1 | XP_005274221.1 | 2 - 3 | 271 | 124 | |
XM_005274165.4 | XP_005274222.1 | 3 - 4 | 296 | 124 | |
XM_006718655.3 | XP_006718718.1 | 3 - 4 | 304 | 124 | |
XM_006718656.3 | XP_006718719.1 | 2 - 3 | 255 | 124 | |
XM_006718657.1 | XP_006718720.1 | 2 - 3 | 237 | 124 | |
XM_017018108.1 | XP_016873597.1 | 3 - 4 | 318 | 124 | |
GenBank mRNA | AB016869.1 | - | 3 - 4 | 295 | 124 |
AB019245.1 | - | 3 - 4 | 293 | 121 | |
AB208827.1 | - | 1 - 2 | 737 | 124 | |
AF076931.1 | - | 3 - 4 | 276 | 124 | |
AF099739.1 | - | 3 - 4 | 280 | 124 | |
AK297527.1 | - | 3 - 4 | 289 | 124 | |
BC000094.2 | - | 3 - 4 | 296 | 124 | |
BC136564.1 | - | 3 - 4 | 297 | 124 | |
BX337895.2 | - | 3 - 4 | 326 | 124 |