The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318934.1 | NP_001305863.1 | 2 - 3 | 443 | 135 |
NM_001318935.1 | NP_001305864.1 | 2 - 3 | 443 | 135 | |
NM_003071.3 | NP_003062.2 | 2 - 3 | 443 | 135 | |
NM_139048.2 | NP_620636.1 | 2 - 3 | 443 | 135 | |
XM_011513091.1 | XP_011511393.1 | 2 - 3 | 443 | 135 | |
XM_011513092.1 | XP_011511394.1 | 2 - 3 | 443 | 135 | |
XM_011513093.1 | XP_011511395.1 | 2 - 3 | 443 | 135 | |
XM_017007078.1 | XP_016862567.1 | 2 - 3 | 422 | 135 | |
XM_017007079.1 | XP_016862568.1 | 2 - 3 | 443 | 135 | |
GenBank mRNA | AB209052.1 | - | 2 - 3 | 225 | 135 |
AK291231.1 | - | 2 - 3 | 390 | 135 | |
BC005260.1 | - | 2 - 3 | 407 | 135 | |
BC015498.1 | - | 2 - 3 | 399 | 135 | |
BC030976.1 | - | 2 - 3 | 443 | 135 | |
BC044659.1 | - | 2 - 3 | 411 | 135 | |
L34673.1 | - | 2 - 3 | 402 | 135 | |
S64671.1 | - | 1 - 2 | 114 | 135 |