Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
Chromosome Location
Chr.19: 1107634 - 1174283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100122.1 NP_001093592.1 21 - 22 2716 91
NM_014963.2 NP_055778.2 24 - 25 3048 91
XM_011527804.2 XP_011526106.1 24 - 25 3021 91
GenBank mRNA AB023180.1 - 24 - 25 3025 91
AK074102.1 - 20 - 21 2508 91
AK125139.1 - 21 - 22 2663 91
AK292407.1 - 24 - 25 3048 91
BC106021.1 - 1 - 2 120 91

View Full Product Details