The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100122.1 | NP_001093592.1 | 4 - 5 | 538 | 91 |
NM_014963.2 | NP_055778.2 | 7 - 8 | 870 | 91 | |
XM_011527804.2 | XP_011526106.1 | 7 - 8 | 843 | 91 | |
GenBank mRNA | AB023180.1 | - | 7 - 8 | 847 | 91 |
AK074102.1 | - | 3 - 4 | 330 | 91 | |
AK125139.1 | - | 4 - 5 | 485 | 91 | |
AK292407.1 | - | 7 - 8 | 870 | 91 | |
AK296849.1 | - | 4 - 5 | 485 | 91 | |
AK300981.1 | - | 8 - 9 | 810 | 91 | |
AK309167.1 | - | 4 - 5 | 485 | 91 |