The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006786.3 | NP_006777.1 | 2 - 3 | 253 | 145 |
NM_021995.2 | NP_068835.1 | 3 - 4 | 317 | 145 | |
XM_011540537.2 | XP_011538839.1 | 4 - 5 | 766 | 145 | |
XM_011540538.1 | XP_011538840.1 | 3 - 4 | 411 | 145 | |
XM_017000116.1 | XP_016855605.1 | 4 - 5 | 766 | 145 | |
XM_017000117.1 | XP_016855606.1 | 3 - 4 | 411 | 145 | |
XM_017000118.1 | XP_016855607.1 | 2 - 3 | 218 | 145 | |
GenBank mRNA | AF104118.1 | - | 2 - 3 | 244 | 145 |
AF140630.1 | - | 3 - 4 | 257 | 145 | |
AY358375.1 | - | 2 - 3 | 253 | 145 | |
BC126443.1 | - | 2 - 3 | 294 | 145 | |
BC171734.1 | - | 2 - 3 | 294 | 145 |