The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001759.3 | NP_001750.1 | 2 - 3 | 718 | 64 |
GenBank mRNA | AK223577.1 | - | 2 - 3 | 691 | 64 |
AK291146.1 | - | 2 - 3 | 656 | 64 | |
BC010958.1 | - | 2 - 3 | 661 | 64 | |
BC089384.1 | - | 2 - 3 | 670 | 64 | |
BT019847.1 | - | 2 - 3 | 413 | 64 | |
D13639.1 | - | 2 - 3 | 682 | 64 | |
M90813.1 | - | 2 - 3 | 434 | 64 | |
X68452.1 | - | 2 - 3 | 568 | 64 |