Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTBS
Entrez Gene ID
1486
Gene Name
chitobiase
Gene Aliases
CTB
Chromosome Location
Chr.1: 84553121 - 84574480 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004388.2 NP_004379.1 6 - 7 1022 136
GenBank mRNA AK075423.1 - 6 - 7 1286 136
AK298703.1 - 6 - 7 973 136
AK314640.1 - 6 - 7 988 136
BC024007.2 - 5 - 6 804 136
BC096752.1 - 5 - 6 804 136
BC126333.1 - 6 - 7 998 136
BC126335.1 - 6 - 7 998 136
M95767.1 - 6 - 7 957 136

View Full Product Details