Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFAIP8L1
Entrez Gene ID
126282
Gene Name
TNF alpha induced protein 8 like 1
Gene Aliases
TIPE1
Chromosome Location
Chr.19: 4639505 - 4655568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167942.1 NP_001161414.1 1 - 2 140 69

View Full Product Details