The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130016.2 | NP_001123488.1 | 4 - 5 | 946 | 90 |
NM_001130017.2 | NP_001123489.1 | 4 - 5 | 962 | 90 | |
NM_001179.5 | NP_001170.2 | 4 - 5 | 946 | 90 | |
XM_005262997.1 | XP_005263054.1 | 3 - 4 | 821 | 90 | |
XM_005262999.1 | XP_005263056.1 | 3 - 4 | 821 | 90 | |
XM_005263003.1 | XP_005263060.1 | 3 - 4 | 821 | 90 | |
XM_005263004.1 | XP_005263061.1 | 3 - 4 | 821 | 90 | |
XM_006714220.1 | XP_006714283.1 | 3 - 4 | 821 | 90 | |
XM_011531971.1 | XP_011530273.1 | 3 - 4 | 821 | 90 | |
XM_017008206.1 | XP_016863695.1 | 3 - 4 | 821 | 90 | |
XM_017008207.1 | XP_016863696.1 | 3 - 4 | 821 | 90 | |
XM_017008208.1 | XP_016863697.1 | 5 - 6 | 1334 | 90 | |
XM_017008209.1 | XP_016863698.1 | 3 - 4 | 821 | 90 | |
XM_017008210.1 | XP_016863699.1 | 5 - 6 | 1332 | 90 | |
GenBank mRNA | AK225860.1 | - | 4 - 5 | 942 | 90 |
AK295305.1 | - | 4 - 5 | 928 | 90 | |
AK308810.1 | - | 4 - 5 | 928 | 90 | |
AY550341.1 | - | 2 - 3 | 68 | 90 | |
AY550342.1 | - | 2 - 3 | 68 | 90 | |
AY550344.1 | - | 2 - 3 | 68 | 90 | |
AY550345.1 | - | 2 - 3 | 68 | 90 | |
AY550346.1 | - | 2 - 3 | 68 | 90 | |
BC008397.1 | - | 4 - 5 | 933 | 90 | |
BC017913.1 | - | 4 - 5 | 933 | 90 | |
BT007249.1 | - | 3 - 4 | 812 | 90 | |
CR541664.1 | - | 3 - 4 | 812 | 90 | |
U47054.1 | - | 4 - 5 | 842 | 90 |