Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRT4
Entrez Gene ID
401399
Gene Name
proline rich transmembrane protein 4
Gene Aliases
-
Chromosome Location
Chr.7: 128350325 - 128362067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114726.2 NP_001108198.2 4 - 5 1081 80
NM_001174164.1 NP_001167635.1 5 - 6 1198 80
XM_005250342.3 XP_005250399.1 4 - 5 1198 80
XM_005250343.3 XP_005250400.1 4 - 5 1107 80
XM_006715985.3 XP_006716048.1 4 - 5 1211 80
XM_017012205.1 XP_016867694.1 5 - 6 2017 80
XM_017012206.1 XP_016867695.1 4 - 5 1990 80
GenBank mRNA BX327107.2 - 3 - 4 363 80

View Full Product Details