Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ8A
Entrez Gene ID
56997
Gene Name
coenzyme Q8A
Gene Aliases
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9
Chromosome Location
Chr.1: 226939339 - 226987545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020247.4 NP_064632.2 6 - 7 1027 75
XM_005273201.1 XP_005273258.1 6 - 7 923 75
XM_011544238.1 XP_011542540.1 7 - 8 1119 75
XM_011544239.2 XP_011542541.1 8 - 9 1142 75
XM_011544240.2 XP_011542542.1 6 - 7 936 75
XM_011544241.2 XP_011542543.1 6 - 7 898 75
XM_017001852.1 XP_016857341.1 6 - 7 1117 75
GenBank mRNA AB073905.1 - 6 - 7 950 75
AJ278126.1 - 2 - 3 245 75
AK090494.1 - 6 - 7 764 75
AK092784.1 - 6 - 7 750 75
AK293571.1 - 7 - 8 924 75
AK297735.1 - 3 - 4 306 75
AK300537.1 - 3 - 4 257 75
AK301012.1 - 8 - 9 948 75
BC005171.2 - 6 - 7 976 75
BC129930.1 - 12 - 13 1869 75
BX648860.1 - 6 - 7 848 75

View Full Product Details