Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPATA6L
- Entrez Gene ID
-
55064
- Gene Name
- spermatogenesis associated 6 like
- Gene Aliases
-
C9orf68, bA6J24.2
- Chromosome Location
-
Chr.9: 4540092 - 4666674 on Build GRCh38
View Full Product Details