The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000508.4 | NP_000499.1 | 4 - 5 | 588 | 105 |
NM_021871.3 | NP_068657.1 | 4 - 5 | 588 | 105 | |
GenBank mRNA | AK290559.1 | - | 4 - 5 | 572 | 105 |
BC070246.1 | - | 4 - 5 | 567 | 105 | |
BC098280.1 | - | 4 - 5 | 532 | 105 | |
BC099706.1 | - | 4 - 5 | 532 | 105 | |
BC099720.1 | - | 4 - 5 | 532 | 105 | |
BC101935.1 | - | 4 - 5 | 532 | 105 | |
BC105803.1 | - | 4 - 5 | 571 | 105 | |
CB122305.1 | - | 1 - 2 | 155 | 105 | |
CN393696.1 | - | 4 - 5 | 605 | 105 | |
KU178063.1 | - | 4 - 5 | 509 | 105 | |
M58569.1 | - | 4 - 5 | 538 | 105 |