Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB3A
Entrez Gene ID
5864
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.19: 18196799 - 18204064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011528164.1 XP_011526466.1 1 - 2 165 66

View Full Product Details