Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB3A
Entrez Gene ID
5864
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.19: 18196799 - 18204064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002866.4 NP_002857.1 3 - 4 516 69
XM_011528164.1 XP_011526466.1 3 - 4 513 69
GenBank mRNA AF254795.1 - 3 - 4 476 69
AF498931.1 - 2 - 3 347 69
AK289559.1 - 3 - 4 494 69
AK293467.1 - - 490 69
BC011782.2 - 3 - 4 481 69
M28210.1 - 3 - 4 386 69
X13900.1 - 3 - 4 386 69

View Full Product Details