The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 10 - 11 | 1767 | 125 |
NM_000642.2 | NP_000633.2 | 10 - 11 | 1689 | 125 | |
NM_000643.2 | NP_000634.2 | 10 - 11 | 1487 | 125 | |
NM_000644.2 | NP_000635.2 | 10 - 11 | 1427 | 125 | |
NM_000645.2 | NP_000636.2 | 8 - 9 | 1500 | 125 | |
NM_000646.2 | NP_000637.2 | 10 - 11 | 1500 | 125 | |
XM_005270557.2 | XP_005270614.1 | 10 - 11 | 1382 | 125 | |
GenBank mRNA | AB208867.1 | - | 9 - 10 | 1293 | 125 |
AB621804.1 | - | 10 - 11 | 1637 | 125 | |
BC078663.1 | - | 10 - 11 | 1409 | 125 | |
M85168.1 | - | 8 - 9 | 1502 | 125 | |
U84007.1 | - | 10 - 11 | 1689 | 125 | |
U84008.1 | - | 10 - 11 | 1427 | 125 | |
U84009.1 | - | 10 - 11 | 1487 | 125 | |
U84010.1 | - | 10 - 11 | 1767 | 125 | |
U84011.1 | - | 10 - 11 | 1500 | 125 |