The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 23 - 24 | 3559 | 89 |
NM_000642.2 | NP_000633.2 | 23 - 24 | 3481 | 89 | |
NM_000643.2 | NP_000634.2 | 23 - 24 | 3279 | 89 | |
NM_000644.2 | NP_000635.2 | 23 - 24 | 3219 | 89 | |
NM_000645.2 | NP_000636.2 | 21 - 22 | 3292 | 89 | |
NM_000646.2 | NP_000637.2 | 23 - 24 | 3292 | 89 | |
XM_005270557.2 | XP_005270614.1 | 23 - 24 | 3174 | 89 | |
XM_017000501.1 | XP_016855990.1 | 11 - 12 | 1408 | 89 | |
GenBank mRNA | AB208867.1 | - | 22 - 23 | 3085 | 89 |
AB621804.1 | - | 23 - 24 | 3429 | 89 | |
BC078663.1 | - | 23 - 24 | 3201 | 89 | |
M85168.1 | - | 21 - 22 | 3294 | 89 | |
U84007.1 | - | 23 - 24 | 3481 | 89 | |
U84008.1 | - | 23 - 24 | 3219 | 89 | |
U84009.1 | - | 23 - 24 | 3279 | 89 | |
U84010.1 | - | 23 - 24 | 3559 | 89 | |
U84011.1 | - | 23 - 24 | 3292 | 89 |