Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBQLN1
Entrez Gene ID
29979
Gene Name
ubiquilin 1
Gene Aliases
DA41, DSK2, PLIC-1, UBQN, XDRP1
Chromosome Location
Chr.9: 83659963 - 83708253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013438.4 NP_038466.2 2 - 3 906 77
NM_053067.2 NP_444295.1 2 - 3 906 77
XM_005251948.2 XP_005252005.1 2 - 3 636 77
GenBank mRNA AB035275.1 - 2 - 3 502 77
AB209468.1 - 1 - 2 50 77
AF176069.3 - 2 - 3 332 77
AF293384.1 - 2 - 3 332 77
AK027556.1 - 2 - 3 605 77
AK027576.1 - 2 - 3 596 77
AK074535.1 - 2 - 3 858 77
AL136643.1 - 2 - 3 610 77
BC010066.2 - 2 - 3 586 77
BC039294.1 - 2 - 3 597 77

View Full Product Details