Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
Chromosome Location
Chr.9: 127815012 - 127854773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000118.3 NP_000109.1 5 - 6 1108 84
NM_001114753.2 NP_001108225.1 5 - 6 1108 84
NM_001278138.1 NP_001265067.1 5 - 6 848 84
GenBank mRNA AK222669.1 - 5 - 6 1021 84
AK290389.1 - 5 - 6 992 84
AK301171.1 - 5 - 6 848 84
AK302455.1 - 2 - 3 220 84
BC014271.2 - 5 - 6 842 84
BT006872.1 - 5 - 6 690 84
J05481.1 - 5 - 6 651 84
X72012.1 - 5 - 6 971 84

View Full Product Details