The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000118.3 | NP_000109.1 | 9 - 10 | 1689 | 63 |
NM_001114753.2 | NP_001108225.1 | 9 - 10 | 1689 | 63 | |
NM_001278138.1 | NP_001265067.1 | 9 - 10 | 1429 | 63 | |
GenBank mRNA | AK222669.1 | - | 9 - 10 | 1602 | 63 |
AK223044.1 | - | 2 - 3 | 252 | 63 | |
AK290389.1 | - | 9 - 10 | 1573 | 63 | |
AK300156.1 | - | 4 - 5 | 406 | 63 | |
AK301171.1 | - | 9 - 10 | 1429 | 63 | |
AK302455.1 | - | 6 - 7 | 801 | 63 | |
BC014271.2 | - | 9 - 10 | 1423 | 63 | |
BT006872.1 | - | 9 - 10 | 1271 | 63 | |
J05481.1 | - | 9 - 10 | 1232 | 63 | |
X72012.1 | - | 9 - 10 | 1552 | 63 |