Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLANA
Entrez Gene ID
2315
Gene Name
melan-A
Gene Aliases
MART-1, MART1
Chromosome Location
Chr.9: 5890909 - 5909822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005511.1 NP_005502.1 3 - 4 226 78
GenBank mRNA AK312149.1 - 3 - 4 242 78
BC014423.1 - 3 - 4 251 78
CR450299.1 - 2 - 3 173 78
U06452.1 - 3 - 4 226 78
U06654.1 - 3 - 4 333 78

View Full Product Details